missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PP1 alpha (Thr320) Recombinant Rabbit Monoclonal Antibody (8C7U9)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | Phospho-PP1 alpha (Thr320) |
|---|---|
| Clone | 8C7U9 |
| Concentration | 0.071 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282849
|
Invitrogen™
MA558249 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283833
![]() |
Invitrogen™
MA558250 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: PITPP.
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| Phospho-PP1 alpha (Thr320) | |
| 0.071 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P62136, P62137, P62138 | |
| 19045, 24668, 5499 | |
| A synthetic phosphorylated peptide around T320 of human PPP1CA (NP_002699.1). | |
| Antibody |
| 8C7U9 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PPP1CA | |
| 96A; CG6593; CG6593-PA; CG6593-PB; dism2; Dmel\CG6593; Dmel_CG6593; DmPp1-96A; GADD34; LD14639p; MGC15877; MGC1674; PP1; PP1 96A; PP1 alpha96a; Pp1-96A; PP1A; PP-1A; PP1A-96A; PP1alpha; PP1alpha[[1]]; PP1alpha96A; pp1alpha-96A; Pp1alpha-96A-PA; Pp1alpha-96A-PB; PP1c; PP1c 96A; PPP1A; Ppp1c; Ppp1ca; protein phosphatase 1; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1, catalytic subunit, alpha isoform; protein phosphatase 1, catalytic subunit, alpha isozyme; Protein phosphatase 1alpha at 96A; protein phosphatase type 1 alpha catalytic subunit; Protein phosphatase type 1 alpha, catalytic subunit; protein phosphatase-1d; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; Serine/threonine-protein phosphatase alpha-1 isoform; Serine/threonine-protein phosphatase PP1-alpha catalytic subunit; testicular tissue protein Li 155 | |
| PPP1CA | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title