Learn More
Phospho-PKR (Thr446) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537704
Code : NEW
Additional Details : Weight : 0.00910kg
Description
A suggested positive control for Western blot is K562 cells; suggested positive control for IHC is human colon carcinoma; suggested positive control for ICC/IF is Hela cells.
Phospho-PKR (Thr446) Polyclonal antibody specifically detects Phospho-PKR (Thr446) in Human samples. It is validated for Western Blot.Specifications
Phospho-PKR (Thr446) | |
Polyclonal | |
Unconjugated | |
EIF2AK2 | |
2310047A08Rik; 4732414G15Rik; AI467567; AI747578; double stranded RNA activated protein kinase; dsRNA-activated kinase; EC 2.7.11.1; eIF-2 alpha; eIF2a Kinase; eIF-2A protein kinase 2; EIF2AK1; EIF2AK2; eukaryotic translation initiation factor 2 alpha kinase 2; eukaryotic translation initiation factor 2-alpha kinase 2; IFN- type I-induced and dsRNA-activated kinase; IFN-induced and double-stranded RNA-activated kinase; interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; interferon-inducible RNA-dependent protein kinase; MGC126524; OTTHUMP00000201320; P1/eIF-2A protein kinase; p68 kinase; PKR; PPP1R83; Prkr; protein kinase R; protein kinase RNA-activated; protein kinase, interferon inducible double stranded RNA dependent; Protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; serine/threonine-protein kinase TIK; T-cell viral integration site; Tik; Tyrosine-protein kinase EIF2A | |
Rabbit | |
Affinity chromatography | |
RUO | |
5610 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P19525 | |
EIF2AK2 | |
Peptide sequence around phosphorylation site of threonine 446 (K-R-T(p)-R-S) derived from Human PKR. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |