missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PKC Pan (Ser660) Recombinant Rabbit Monoclonal Antibody (24GB5450)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557164
This item is not returnable.
View return policy
Description
Phospho-PKC Pan (Ser660) Recombinant Monoclonal Antibody for Western Blot, Flow
Members of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differ among tissues and PKC family members exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca2+.
Specifications
| Phospho-PKC Pan (Ser660) | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| F1M7Y5, P05129, P05696, P05771, P09215, P09216, P09217, P16054, P20444, P23298, P24723, P28867, P41743, P63318, P63319, P68403, P68404, Q02111, Q02156, Q02956, Q04759, Q05513, Q05655, Q62074, Q64617, Q9WTQ0 | |
| Prkca, PRKCB, PRKCD, PRKCE, PRKCG, Prkch, Prkci, PRKCQ, PRKCZ | |
| A synthesized peptide derived from human Phospho-PKC (Ser660) (aa 654-664). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5450 | |
| Unconjugated | |
| PRKCB | |
| 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; 2310021H13Rik; 5830406C15Rik; A130035A12Rik; A130082F03Rik; AAG6; aging-associated gene 6; AI098070; AI385711; AI427505; AI875142; ALPS3; aPKClambda; aPKC-lambda/iota; aPKCzeta; atypical protein kinase C; atypical protein kinase C-lambda/iota; AW494342; C80388; CVID9; D14Ertd420e; DXS1179E; MAY1; mKIAA4165; nPKC-delta; nPKC-epsilon; nPKC-eta; nPKC-iota; nPKC-theta; nPKC-zeta; PKC; PKC theta; PKC zeta; PKC[d]; PKC[e]; PKC-0; PKC2; Pkca; PKC-A; PKC-alpha; Pkcb; PKC-B; PKC-beta; Pkcc; PKCD; PKCdelta; Pkce; Pkcea; PKCepsilon; PKCG; PKCgamma; PKC-gamma; Pkch; PKCI; PKCL; PKC-L; PKClambda; Pkcq; PKCtheta; PKC-theta; Pkcz; PKC-ZETA; PRKACA; Prkc; PRKCA; Prkcb; Prkcb1; Prkcb2; Prkcc; Prkcd; Prkce; PRKCG; Prkch; PRKCI; PRKCL; PRKC-lambda/iota; Prkcq; PRKCT; Prkcz; protein kinase C alpha; protein kinase C alpha type; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C eta; protein kinase C eta type; protein kinase C gamma; protein kinase C gamma type; protein kinase C iota; protein kinase C iota type; protein kinase C theta; protein kinase C theta type; protein kinase C type I (gamma type); protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, alpha; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C, epsilon; protein kinase C, eta; protein kinase C, gamma; protein kinase C, iota; protein kinase C, lambda; protein kinase C, theta; protein kinase C, zeta; protein kinase C[d]; protein kinase c-alpha; protein kinase C-eta; r14-3-3; R74924; R75156; RATPKCI; SCA14; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; zetaPKC | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170538, 18750, 18751, 18752, 18753, 18754, 18755, 18759, 18761, 18762, 24680, 24681, 25023, 25522, 29340, 5579, 5580, 5581, 5582, 5583, 5584, 5588, 5590, 81749, 84006, 85420 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction