missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PKC lambda/iota (Thr557, Thr564) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ 44968G
This item is not returnable.
View return policy
Description
Phospho-PKC lambda/iota (Thr557, Thr564) Polyclonal Antibody for Western Blot
The PKC family of serine/threonine kinases, including PRKCI (PKC iota), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X.
Specifications
| Phospho-PKC lambda/iota (Thr557, Thr564) | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
| P41743, Q62074 | |
| PRKCI | |
| The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human PKCi that contains threonine 557 and 564 (phosphorylated). The sequence is conserved in mouse PKCi and human PKCl. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PRKCI | |
| 2310021H13Rik; AI427505; aPKClambda; aPKC-lambda/iota; atypical protein kinase C-lambda/iota; DXS1179E; hypothetical protein; mKIAA4165; nPKC-iota; Pkci; Pkcl; PKClambda; PRKCI; Prkcl; PRKC-lambda/iota; protein kinase C iota; protein kinase C iota type; protein kinase C, iota; protein kinase C, lambda | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18759, 5584 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction