missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PKC delta (Thr507) Rabbit, anti-Human, Polyclonal, Proteintech

Product Code. 18059635 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18059635 20 μL 20µL
18086665 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18059635 Supplier Proteintech Supplier No. 295641AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Phospho-PKC Delta (Thr507) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-PKC Delta (Thr507). This antibody recognizes human antigen. The Phospho-PKC Delta (Thr507) antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen Phospho-PKC delta (Thr507)
Applications Western Blot, ELISA
Concentration 550 μg/mL
Gene Symbols MAY1, PKCD, PRKCD, Protein kinase C delta type, protein kinase C, delta
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 5580
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.