Learn More
Phospho-PKC Alpha (Thr638) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29123-1-AP-20UL
Description
The Phospho-PKC Alpha (Thr638) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-PKC Alpha (Thr638). This antibody recognizes human, mouse antigen. The Phospho-PKC Alpha (Thr638) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-PKC Alpha (Thr638) | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Western Blot, ELISA | |
| Unconjugated | |
| 20 μL | |
| 5578 | |
| IgG |