missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PIK3CG (Ser1100) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA512894
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and porcine based on sequence homology.
PIK3CG (p110 gamma) is the sole class IB phosphatidylinositol 3-kinase family member, which along with the class IA family members, generates the important second messenger PIP2. However, unlike the class IA enzymes which signal downstream of receptor tyrosine kinases, PIK3CG primarily signals downstream of GPCRs. The gene product is an enzyme that phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring. It is an important modulator of extracellular signals, including those elicited by E-cadherin-mediated cell-cell adhesion, which plays an important role in maintenance of the structural and functional integrity of epithelia. In addition to its role in promoting assembly of adherens junctions, the protein is thought to play a pivotal role in the regulation of cytotoxicity in NK cells. The gene is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias.
Specifications
| Phospho-PIK3CG (Ser1100) | |
| Polyclonal | |
| Unconjugated | |
| PIK3CG | |
| 1-phosphatidylinositol 3-kinase; 5830428L06Rik; EGK_14026; p110gamma; p110-gamma; p120-PI3K; phosphatidylinositol 3 kinase gamma, p110 gamma; phosphatidylinositol 3-kinase catalytic 110-kD gamma; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma; Phosphoinositide-3-kinase catalytic gamma polypeptide; phosphoinositide-3-kinase gamma catalytic subunit; phosphoinositide-3-kinase, catalytic, gamma polypeptide; PI3CG; PI3K; pi3kcg; Pi3kg1; PI3Kgamma; PI3K-gamma; PI3-kinase subunit gamma; PIK3; Pik3cg; ptdIns-3-kinase subunit gamma; ptdIns-3-kinase subunit p110-gamma; Serine/threonine protein kinase PIK3CG | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P48736 | |
| PIK3CG | |
| KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S1100 of human PIK3CG. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction