missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Phospholamban (Ser16) Recombinant Rabbit Monoclonal Antibody (HL1710)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546899
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Phospholamban (PLB) is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer.
Specifications
| Phospho-Phospholamban (Ser16) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P26678, P61014 | |
| PLN | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Ser16 of human phospholamban . The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1710 | |
| Unconjugated | |
| PLN | |
| cardiac CMH18; Cardiac phospholamban; CMD1P; CMH18; phospholamban; Plb; Plm; PLN; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18821, 5350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction