missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PERK/EIF2AK3 (Ser719) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£109.00 - £249.00
Specifications
| Antigen | PERK/EIF2AK3 (Ser719) |
|---|---|
| Concentration | 250 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233269
|
Proteintech
29546-1-AP-100UL |
100 μL |
£280.00 £249.00 / 100µL Save £31.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235175
|
Proteintech
29546-1-AP-20UL |
20 μL |
£125.00 £109.00 / 20µL Save £16.00 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-PERK/EIF2AK3 (Ser719) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-PERK/EIF2AK3 (Ser719). This antibody recognizes human antigen. The Phospho-PERK/EIF2AK3 (Ser719) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| PERK/EIF2AK3 (Ser719) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp781H1925; HRI; PEK; PERK; WRS | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 250 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 9451 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title