missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PDH E1 Alpha (Ser232) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
£119.00 - £330.00
Specifications
| Antigen | PDH E1 Alpha (Ser232) |
|---|---|
| Clone | 1L22 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236472
|
Proteintech
81491-1-RR-100UL |
100 μL |
£360.00 £330.00 / 100µL Save £30.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233412
|
Proteintech
81491-1-RR-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-PDH E1 Alpha (Ser232) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-PDH E1 Alpha (Ser232). This antibody recognizes human antigen. The Phospho-PDH E1 Alpha (Ser232) antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| PDH E1 Alpha (Ser232) | |
| 500 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 5160 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1L22 | |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| PDHA; PDHCE1A; PHE1A | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel