missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PDGFRB (Tyr751) Monoclonal Antibody (G.13.6)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514823
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
PDGFRB is a receptor tyrosine kinase and has been implicated in atherogenesis and in cell transformation by the SIS oncogene. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. The gene is flanked on chromosome 5 by the genes for granulocyte-macrophage colony-stimulating factor and macrophage-colony stimulating factor receptor; all three genes may be implicated in the 5-q syndrome. A translocation between chromosomes 5 and 12, that fuses PDGFRB (CD140b) to that of the translocation, ETV6, leukemia gene, results in chronic myeloproliferative disorder with eosinophilia.
Specifications
| Phospho-PDGFRB (Tyr751) | |
| Monoclonal | |
| Unconjugated | |
| Pdgfrb | |
| AI528809; Beta platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140B; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF beta chain; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; PDGFRB; PDGF-R-beta; PDGFR-beta; PENTT; platelet derived growth factor receptor beta; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18596, 24629, 5159 | |
| -20°C | |
| Liquid |
| Western Blot | |
| G.13.6 | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| P05622, P09619, Q05030 | |
| Pdgfrb | |
| Synthetic phosphopeptide corresponding to residues surrounding pTyr751 of human PDGF receptor β. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction