missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PDCD5 (Ser119) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105376
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of PDCD5 only when phosphorylated at S119.
Programmed cell death 5 (PDCD5), a human apoptosis-related protein, is thought to play an early and universal role in apoptosis. PDCD5 is widely expressed and is upregulated in cells undergoing apoptosis, where it translocates rapidly from the cytoplasm to the nucleus. PDCD5 has a compact core structure of low flexibility with two mobile alpha-helices at N-terminal and a flexible unstructured C-terminal region. The charged residues are crucial for the ability of apoptosis-promoting and cell translocation of the protein. PDCD5 can facilitate apoptosis and enhance TAJ/TROY-induced paraptosis-like cell death. PDCD5 may play a dual role in the Tip60 pathway. It interacts with Tip60 and functions as a Tip60 co-activator to promote apoptosis. The nucleotide polymorphisms in the 5'-upstream region of PDCD5 affect promoter activity and the susceptibility of a Chinese population to develop chronic myelogenous leukemia and may represent a novel tumor suppressor gene influencing lung cancer.
Specifications
| Phospho-PDCD5 (Ser119) | |
| Polyclonal | |
| Unconjugated | |
| PDCD5 | |
| 2200003D22Rik; AA408513; PDCD5; programmed cell death 5; programmed cell death protein 5; Protein TFAR19; TF1 cell apoptosis-related gene 19; TF-1 cell apoptosis-related protein 19; Tfar19; TFAR19 novel apoptosis-related | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 292814, 56330, 9141 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14737, P56812 | |
| PDCD5 | |
| A synthesized peptide derived from human PDCD5(Accession O14737), corresponding to amino acid residues around phosphorylated Ser119. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction