missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PBK (Thr9) Recombinant Rabbit Monoclonal Antibody (23GB4910), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Phospho-PBK (Thr9) |
|---|---|
| Clone | 23GB4910 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087981
![]() |
Invitrogen™
MA553824 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087608
|
Invitrogen™
MA553825 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-PBK (Thr9) Recombinant Monoclonal antibody specifically detects Phospho-PBK (Thr9) in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Phospho-PBK (Thr9) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96KB5 | |
| 55872 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4910 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2810434B10Rik; AW538537; Cancer/testis antigen 84; CT84; D14Ertd732e; epididymis luminal protein 164; HEL164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PBK; PDZ binding kinase; PDZ-binding kinase; serine/threonine protein kinase; spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase; TOPK | |
| A synthesized peptide derived from human Phospho-PBK/TOPK (Thr9) (2-16AA). | |
| Primary | |
| -20°C | |
| PBK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title