missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PBK (Thr9) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104630
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of PBK/TOPK only when phosphorylated at Thr9.
PBK (TOPK) is a serine/threonine kinase that is related to the dual specific mitogen-activated protein kinase (MAPKK). The kinase is up-regulated in a variety of hematological malignancies, participates in activating lymphoid cells, and supports testicular function (spermatogenesis). Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
Specifications
| Phospho-PBK (Thr9) | |
| Polyclonal | |
| Unconjugated | |
| PBK | |
| 2810434B10Rik; AW538537; Cancer/testis antigen 84; CT84; D14Ertd732e; epididymis luminal protein 164; HEL164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PBK; PDZ binding kinase; PDZ-binding kinase; serine/threonine protein kinase; spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase; TOPK | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 52033, 55872 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96KB5, Q9JJ78 | |
| PBK | |
| A synthesized peptide derived from human PBK(Accession Q96KB5), corresponding to amino acid residues around phosphorylated Thr9. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction