missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Paxillin (Tyr118) Recombinant Rabbit Monoclonal Antibody (1F11)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA533194
This item is not returnable.
View return policy
Description
Phospho-Paxillin (Tyr118) Recombinant Monoclonal Antibody for Western Blot, ELISA
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
Specifications
| Phospho-Paxillin (Tyr118) | |
| Recombinant Monoclonal | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49023 | |
| PXN | |
| A synthesized peptide derived from human Phospho-PXN (Y118). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1F11 | |
| Unconjugated | |
| PXN | |
| adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 | |
| Rabbit | |
| Antigen affinity chromatography, Protein G | |
| RUO | |
| 5829 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction