missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PAK4/6/7 (Ser474) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536865
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 72 kDa (PAK4), 82 kDa (PAK6) and 90 (PAK7). Purity is >95% by SDS-PAGE.
PAK4 is highly expressed in prostate, testis and colon. PAK4 interacts tightly with GTP-bound but not GDP-bound CDC42 and weakly with RAC. PAK4 phosphorylates and autophosphorylates and also activates the JNK pathway. PAK5, a member of the PAK family of protein kinases contains a CDC42/Rac1 interactive binding (CRIB) motif at the N-terminus and a Ste20-like kinase domain at the C-terminus. PAK5 preferentially binds to CDC42 in the presence of GTP and the CRIB motif is essential for this interaction. PAK6 protein cotranslocates into the nucleus with androgen receptor, which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development, in response to androgen. PAK6 was initially identified as an androgen receptor in a yeast two hybrid screen and was found to be highly expressed in testis and prostate tissues. Later experiments have shown it to be activated by MAP kinase 6 and p38 MAP kinase, suggesting that PAK6 may play a role in the cellular response to stress-related signals.
Specifications
| Phospho-PAK4/6/7 (Ser474) | |
| Polyclonal | |
| Unconjugated | |
| Pak4 | |
| 2900083L08Rik; 4732456M09; 5730488L07Rik; 6430627N20; AW555722; KIAA1142; KIAA1264; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (CDKN1A)-activated kinase 6; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 4; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 6; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 6; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 4; p21(CDKN1A)-activated kinase 6; p21(CDKN1A)-activated kinase 7; p21-activated kinase 4; p21-activated kinase 5; p21-activated kinase 6; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 4/5/6; PAK 4/6/7; PAK4; PAK-4; PAK4/5/6; PAK5; PAK-5; PAK6; PAK-6; Pak7; PAK-7; protein kinase PAK5; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 6; serine/threonine-protein kinase PAK 6-like; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10298, 56924, 57144 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O96013, Q9NQU5, Q9P286 | |
| Pak4, PAK5, PAK6 | |
| Synthetic phosphopeptide derived from human PAK4 around the phosphorylation site of Serine 474. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction