missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PAK4/6/7 (Ser474) Recombinant Rabbit Monoclonal Antibody (1H7L3)
Phospho-PAK4/6/7 (Ser474) Recombinant Rabbit Monoclonal Antibody (1H7L3)
Brand: Invitrogen 702663
This item is not returnable.
View return policy
Description
Phospho-PAK4/6/7 (Ser474) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-PAK4/6/7 (Ser474) | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O96013, Q9NQU5, Q9P286 | |
| Pak4, PAK5, PAK6 | |
| Peptide corresponding to human Phospho-PAK4/PAK5/PAK6 (Ser474) (aa472-aa480). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1H7L3 | |
| Unconjugated | |
| Pak4 | |
| 2900083L08Rik; 4732456M09; 5730488L07Rik; 6430627N20; AW555722; KIAA1142; KIAA1264; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (CDKN1A)-activated kinase 6; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 4; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 6; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 6; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 4; p21(CDKN1A)-activated kinase 6; p21(CDKN1A)-activated kinase 7; p21-activated kinase 4; p21-activated kinase 5; p21-activated kinase 6; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 4/5/6; PAK 4/6/7; PAK4; PAK-4; PAK4/5/6; PAK5; PAK-5; PAK6; PAK-6; Pak7; PAK-7; protein kinase PAK5; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 6; serine/threonine-protein kinase PAK 6-like; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10298, 56924, 57144 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction