missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PAK2 (Ser20) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17148073
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17148073 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17148073 Supplier Invitrogen™ Supplier No. PA5110143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSSTI.

PAK2 (serine/threonine-protein kinase PAK2) is a serine/threonine kinase that plays a role in a variety of different signaling pathways including cytoskeleton regulation, cell motility, cell cycle progression, apoptosis, or proliferation. It acts as a downstream effector of the small GTPases CDC42 and RAC1. Activation by the binding of active CDC42 and RAC1 results in a conformation change and a subsequent autophosphorylation on several serine and/or threonine residues. Full-length PAK2 stimulates cell survival and cell growth. PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PAK2 (Ser20)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Pak2
Gene Accession No. Q13177, Q8CIN4
Gene Alias 5330420P17Rik; A130002K10Rik; AI836325; Alpha-PAK; Beta-PAK; C-t-PAK2; D16Ertd269e; gamma pak; gamma-PAK; mKIAA4182; Oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 2; p21 (RAC1) activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 2; p21-activated kinase 2; p27; p34; p58; p65-PAK; PAK alpha; PAK beta; PAK gamma; PAK1; PAK-1; Pak2; PAK-2; PAK-2p27; PAK-2p34; PAK3; PAK-3; PAK65; PAKgamma; S6/H4 kinase; serine/threonine-protein kinase PAK 2
Gene Symbols Pak2
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S20 of human PAK2 (NP_002568.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224105, 5062
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.