missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PAK1/2/3 (Ser144) Recombinant Rabbit Monoclonal Antibody (1T7A5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538239
This item is not returnable.
View return policy
Description
Positive Samples: NIH/3T3 Immunogen sequence: YMSFT.
In mammals, there are several identified isoforms of p21-Activated Protein Kinases or PAKs: alpha-PAK (also known as PAK-1) and beta-PAK (also known as PAK-3) are mostly brain-specific, while gamma-PAK (also known as PAK-2) is expressed ubiquitously (Jakobi et al., 2003). Mutations of the gene coding for PAK-3 are associated wit X-linked mental retardation and recent work indicates that PAK-3 is a key regulator of synapse formation and plasticity in the hippocampus (Boda et al., 2004). PAK-3 is thought to play a key role in regulation of cell shape and motility as well as cell death (Jakobi et al., 2003; Walter et al., 1998). Autophosphorylation of Thr402 in the protein has been found to be essential for activation of PAK (Jakobi et al., 2000).
Specifications
| Phospho-PAK1/2/3 (Ser144) | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75914, O88643, P35465, Q13153, Q13177, Q61036, Q62829, Q64303, Q8CIN4 | |
| Pak1, PAK2, PAK3 | |
| A synthetic phosphorylated peptide around S144 of human PAK1PAK1 (Q13153). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1T7A5 | |
| Unconjugated | |
| PAK3 | |
| 5330420P17Rik; A130002K10Rik; activated protein kinase alpha; ADRB2; adriamycin resistance-associated; AI836325; Alpha-PAK; ARA; AW045634; Beta-PAK; bPAK; CDC42/RAC effector kinase PAK-A; CDC42/RAC effector kinase PAK-B; CDKN1A; C-t-PAK2; D16Ertd269e; EC 2.7.11.1; gamma pak; gamma-PAK; hPAK3; kinase PAK1; mKIAA4182; MRX30; MRX47; oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 1; p21 (CDKN1A)-activated kinase 2; p21 (CDKN1A)-activated kinase 3; p21 (RAC1) activated kinase 1; p21 (RAC1) activated kinase 2; p21 (RAC1) activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 1; p21 protein (Cdc42/Rac)-activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 3; p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast); p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p21/Cdc42/Rac1-activated kinase 3 (yeast Ste20-related); p21-activated kinase 1; p21-activated kinase 2; p21-activated kinase 3; p27; p34; p58; p65-PAK; p68-PAK; PAK 1; PAK alpha; PAK beta; PAK gamma; Pak1; PAK-1; Pak2; PAK-2; PAK-2p27; PAK-2p34; PAK3; PAK-3; PAK3beta; PAK65; Pak65alpha; Pak65beta; Paka; PAKalpha; Pakb; PAKgamma; Protein kinase MUK2; S6/H4 kinase; Serine/threonine-protein kinase PAK 1; serine/threonine-protein kinase PAK 2; Serine/threonine-protein kinase PAK 3; STE20 homolog, yeast; Stk4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18479, 18481, 224105, 29431, 29432, 29433, 5058, 5062, 5063 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction