missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Phospho-P53 (Ser392) Polyclonal antibody specifically detects Phospho-P53 (Ser392) in Human samples. It is validated for WB, ELISA
Specifications
Specifications
| Antigen | Phospho-P53 (Ser392) |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 260 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| Gene Accession No. | BC005807 |
| Host Species | Rabbit |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?