missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-OXSR1 (Thr185) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103272
This item is not returnable.
View return policy
Beschreibung
Antibody detects endogenous levels of OSR1 only when phosphorylated at Thr185.
OXSR1 serine/threonine kinase or (OSR1) is activated by osmotic stresses and regulates downstream protein kinases such as PAK1 in order to establish the sensor/signaling modules that initiate the cellular response to environmental stress. OXSR1 can be phosphorylated by ATM or ATR during the DNA damage process. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.
Spezifikation
| Phospho-OXSR1 (Thr185) | |
| Polyclonal | |
| Unconjugated | |
| Oxsr1 | |
| 2210022N24Rik; 2810422B09Rik; AI462649; AW209236; C6orf182; CEP57R; KIAA1101; mKIAA1101; Osr1; OTTHUMP00000209062; oxidative stress responsive 1; oxidative stress responsive kinase 1; oxidative stress-responsive 1 protein; oxidative-stress responsive 1; OXSR1; Serine/threonine-protein kinase OSR1 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 108737, 316064, 9943 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95747, Q6P9R2 | |
| Oxsr1 | |
| A synthesized peptide derived from human OXSR1(Accession O95747), corresponding to amino acid residues around phosphorylated Thr185. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur