missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-OPRD1 (Ser363) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540246
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer's patients and in the putamen in Parkinson's disease.
Specifications
| Phospho-OPRD1 (Ser363) | |
| Polyclonal | |
| Unconjugated | |
| OPRD1 | |
| delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18386, 24613, 4985 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32300, P33533, P41143 | |
| OPRD1 | |
| Synthesized peptide derived from human DOR-1 around the phosphorylation site of S363. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction