missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-OCT1 (POU2F1) (Ser385) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5105826
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of POU2F1/OCT1 only when phosphorylated at Ser385.
OCT1 (POU2F1) is a ubiquitously expressed transcription factor that binds to the octamer motif (5'-ATTTGCAT-3') and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR and PGR. In case of human herpes simplex virus (HSV) infection, POU2F1 forms a multiprotein-DNA complex with the viral transactivator protein VP16 and HCFC1 thereby enabling the transcription of the viral immediate early genes.
Specifications
| Phospho-OCT1 (POU2F1) (Ser385) | |
| Polyclonal | |
| Unconjugated | |
| POU2F1 | |
| 2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 171068, 18986, 5451 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14859, P25425, P31503 | |
| POU2F1 | |
| A synthesized peptide derived from human POU2F1(Accession P14859), corresponding to amino acid residues around phosphorylated Ser385. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction