missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NTAL (Tyr136) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564683
This item is not returnable.
View return policy
Description
Phospho-NTAL (Tyr136) Polyclonal Antibody detects endogenous levels of NTAL only when phosphorylated at Tyr136.
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Specifications
| Phospho-NTAL (Tyr136) | |
| Polyclonal | |
| Unconjugated | |
| LAT2 | |
| AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 317676, 56743, 7462 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CGL2, Q9GZY6, Q9JHL0 | |
| LAT2 | |
| A synthesized peptide derived from human LAB (Phospho-Tyr136). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction