missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Nrf2 (Ser344) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105664
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Nrf2 only when phosphorylated at Ser344.
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
Specifications
| Phospho-Nrf2 (Ser344) | |
| Polyclonal | |
| Unconjugated | |
| NFE2L2 | |
| AI194320; E4TF1-60; E4TF1A; HEBP1; Nfe2l2; NF-E2-related factor 2; NFE2-related factor 2; NF-E2-related factor-2; NFT2; NRF2; NRF2A; nuclear factor (erythroid-derived 2)-like 2; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2; Unknown (protein for IMAGE:7945830) | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 18024, 4780, 83619 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54968, Q16236, Q60795 | |
| NFE2L2 | |
| A synthesized peptide derived from human NFE2L2(Accession Q16236), corresponding to amino acid residues around phosphorylated Ser344. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction