missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Niban-like protein (Ser679, Ser683) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595683
This item is not returnable.
View return policy
Description
Antigen Affinity Purified from Pooled Serum.
FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
Specifications
| Phospho-Niban-like protein (Ser679, Ser683) | |
| Polyclonal | |
| Unconjugated | |
| Fam129b | |
| 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 227737, 64855 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5 | |
| Q8R1F1, Q96TA1 | |
| Fam129b | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser679/683 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction