missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NFKB p65 (Ser468) Recombinant Rabbit Monoclonal Antibody (NFKBp65S468-B9), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£301.00 - £600.00
Specifications
| Antigen | Phospho-NFKB p65 (Ser468) |
|---|---|
| Clone | NFKBp65S468-B9 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17147038
|
Invitrogen™
MA537448 |
20 μL |
£301.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17166788
![]() |
Invitrogen™
MA537200 |
200 μL |
£600.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuclear factor kB p65 (NF-kB p65) is encoded by the RELA gene and is present on chromosome 11 in humans. NF-kB P65 is also known as RelA (v-rel avian reticuloendotheliosis viral oncogene homolog A) and belongs to the Rel family of proteins. It is one of the two subunits of NF-kB (Nuclear factor kappa-light-chain-enhancer of activated B cells) that heterodimerizes with the other subunits p50 or p52. However, binding of TNF alpha to its cognate receptor phosphorylates IKK which in turn phosphorylates I kB allowing proteasomal degradation of I kB. It specifically plays a key role in transcription of immunoglobulin k (kappa) gene in mature B-lymphoid cells.Specifications
| Phospho-NFKB p65 (Ser468) | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
| avian reticuloendotheliosis viral (v-rel) oncogene homolog A; EBP-1; I79_021812; MGC131774; NF KB; NFkappaB p65; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NF-kB p65; NF-kB p65 subunit; nfkb rela; NFKB1; nfkb3; NFkB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; putative transcription factor p65 homolog; REL A; rel1; rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3, p65; XELAEV_18022301mg; Xrel1; xrela | |
| RELA | |
| IgG κ | |
| Protein A/G | |
| Antibody |
| NFKBp65S468-B9 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q04206 | |
| 5970 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser536 of human phospho-NFKB p65. | |
| Primary | |
| -20°C | |
| RELA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title