missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NEDD4L (Ser479) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105691
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of NEDD4L only when phosphorylated at Ser479.
NEDD4L is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. NEDD4L inhibits TGF-beta signaling by triggering SMAD2 and TGFR1 ubiquitination and proteasome-dependent degradation. NEDD4L promotes ubiquitination and internalization of various plasma membrane channels such as ENaC, Nav1.2, Nav1.3, Nav1.5, Nav1.7, Nav1.8, Kv1.3, EAAT1 or CLC5. NEDD4L also promotes ubiquitination and degradation of SGK.
Specifications
| Phospho-NEDD4L (Ser479) | |
| Polyclonal | |
| Unconjugated | |
| NEDD4L | |
| 1300012C07Rik; E3 ubiquitin-protein ligase NEDD4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; hNEDD4-2; KIAA0439; NEDD4; NEDD4 like E3 ubiquitin protein ligase; NEDD4.2; Nedd4-2; nedd4b; NEDD4L; NEDL3; neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4b; neural precursor cell expressed, developmentally down-regulated gene 4-like; RSP5; ubiquitin-protein ligase Nedd4-2; ubiquitin-protein ligase Rsp5; Unknown (protein for IMAGE:7986262) | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 23327, 291553 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96PU5 | |
| NEDD4L | |
| A synthesized peptide derived from human NEDD4L(Accession Q96PU5), corresponding to amino acid residues around phosphorylated Ser479. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction