missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NDRG1 (Thr346) Recombinant Rabbit Monoclonal Antibody (NDRG1T346-F5), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536982
This item is not returnable.
View return policy
Description
NDRG1 is a 394 amino acid cytoplasmic protein belonging to the NDRG family. It interacts with APO A-I and A-II and plays a role in HDL-mediated cholesterol transport, stress responses, hormone responses, cell growth and differentiation. Its defect causes Charcot-Marie-Tooth disease type 4D (CMT4D) that is characterized by reduced nerve conduction velocities, segmental demyelination and remyelination with onion bulb formations on nerve biopsy, slowly progressive distal muscle atrophy and weakness, absent deep tendon reflexes and hollow feet. NDRG1 plays a growth inhibitory role and serves as a marker of differentiation. It is expressed in placental membranes, prostate, kidney, small intestine, ovary tissues, colon and the cells that border the lumen.
Specifications
| Phospho-NDRG1 (Thr346) | |
| Recombinant Monoclonal | |
| PE | |
| NDRG1 | |
| CAP43; CMT4D; differentiation-related gene 1 protein; DRG1; DRG-1; GC4; HMSNL; Ndr1; Ndrg1; Ndrl; nickel-specific induction protein Cap43; NMSL; N-myc downstream regulated 1; N-myc downstream regulated gene 1; N-myc downstream-regulated gene 1 protein; Protein Ndr1; protein NDRG1; protein regulated by oxygen-1; PROXY1; Reducing agents and tunicamycin-responsive protein; Rit42; RTP; TARG1; TDD5 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 10397, 17988 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| NDRG1T346-F5 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| Q62433, Q92597 | |
| NDRG1 | |
| A synthetic phospho-peptide corresponding to residues surrounding Thr346 of human phospho NDRG1. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction