missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NACA (Thr159) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564694
This item is not returnable.
View return policy
Description
Phospho-NACA (Thr159) Polyclonal Antibody detects endogenous levels of NACA only when phosphorylated at Thr159.
NACA prevents inappropriate targeting of non-secretory polypeptides to the endoplasmic reticulum (ER). It binds to nascent polypeptide chains as they emerge from the ribosome and blocks their interaction with the signal recognition particle (SRP), which normally targets nascent secretory peptides to the ER. NACA also reduces the inherent affinity of ribosomes for protein translocation sites in the ER membrane (M sites), and may act as a specific coactivator for JUN, binding to DNA and stabilizing the interaction of JUN homodimers with target gene promoters.
Specifications
| Phospho-NACA (Thr159) | |
| Polyclonal | |
| Unconjugated | |
| NACA | |
| AL022831; AL024382; Alpha-NAC; Alpha-NAC, muscle-specific form; alpha-NAC/1.9.2; Gm1878; Hom s 2; HSD48; mKIAA0363; NACA; NACA1; NAC-alpha; nascent polypeptide-associated complex alpha polypeptide; nascent polypeptide-associated complex alpha subunit; nascent polypeptide-associated complex subunit alpha; Nascent polypeptide-associated complex subunit alpha, muscle-specific form; nascent-polypeptide-associated complex alpha polypeptide; skNAC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17938, 288770, 4666 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9PAV3, P70670, Q13765, Q60817 | |
| NACA | |
| A synthesized peptide derived from human NACA (Phospho-Thr159). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction