Learn More
Phospho-MYL2 (Ser15) Recombinant Rabbit Monoclonal Antibody (HL2891)
GREENER_CHOICE
Phospho-MYL2 (Ser15) Recombinant Rabbit Monoclonal Antibody (HL2891)
Brand:  Invitrogen MA556808
Product Code. 30276427
  • £434.00 / 100µL
Description
Phospho-MYL2 (Ser15) Recombinant Monoclonal Antibody for WB, ICC/IF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-MYL2 (Ser15)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P10916, P51667
MYL2
Carrier-protein conjugated synthetic peptide surrounding phospho Ser15 of human Myosin Light Chain 2 (MLC-2v). The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot, Immunocytochemistry
HL2891
Unconjugated
MYL2
cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; CMH10; DKFZp779C0562; hypothetical protein LOC677750; Mlc2; MLC-2; mlc2b; MLC-2s/v; MLC2V; MLC-2v; myl2; myl2b; Mylpc; myosin light chain 2; myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2V; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin regulatory light chain 2b; myosin regulatory light chain ventricular isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light chain 2b, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; myosin, light polypeptide 2b, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; ventricular myosin light chain 2; ventricular myosin regulatory light chain; zgc:136848
Rabbit
Protein A
RUO
17906, 4633
-20°C, Avoid Freeze/Thaw Cycles