missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-mTOR (Ser2448) Recombinant Rabbit Monoclonal Antibody (mTORS2448-E11), PE, Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£185.00 - £463.00

Specifications

Antigen Phospho-mTOR (Ser2448)
Clone mTORS2448-E11
Applications Flow Cytometry
Classification Recombinant Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17176978
View Documents
Invitrogen™
MA537391
10 Tests
£185.00
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17116738
GREENER_CHOICE
View Documents
Invitrogen™
MA537145
100 Tests
£463.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Specifications

Specifications

Phospho-mTOR (Ser2448)
Flow Cytometry
PE
Rabbit
Human, Mouse
P42345, Q9JLN9
2475, 56717
A synthetic phospho-peptide corresponding to residues surrounding Ser2448 of human phospho mTOR.
Primary
4°C
MTOR
mTORS2448-E11
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
MTOR
IgG κ
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.