missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MSK1 (Thr581) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538370
This item is not returnable.
View return policy
Description
MSK-1 is a mitogen and stress activated protein kinase-1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK-1can be activated by ERK1/2 and SAPK2 /p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG-14 in response to mitogen and stress. Similar to RSK, MSK-1 contains two kinase domains (N-term and a C-term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK-1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C-terminal kinase domain of MSK-1 which is essential for the catalytic activity of the N-terminal kinase domain.
Specifications
| Phospho-MSK1 (Thr581) | |
| Polyclonal | |
| Unconjugated | |
| RPS6KA5 | |
| Ribosomal protein S6 kinase alpha-5, S6K-alpha-5, 90 kDa ribosomal protein S6 kinase 5, Nuclear mitogen- and stress-activated protein kinase 1, RSK-like protein kinase, RSKL, RPS6KA5, MSK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 73086, 9252 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75582, Q8C050 | |
| RPS6KA5 | |
| A synthetic phosphopeptide derived from human MSK1 around the phosphorylation site of Thr581 (L-K-TP-P-C) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction