missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MRLC2 (Ser18) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538375
This item is not returnable.
View return policy
Description
The activity of nonmuscle myosin II (see MYH9; MIM 160775) is regulated by phosphorylation of a regulatory light chain, such as MRLC2. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments (Iwasaki et al., 2001 [PubMed 11942626]).[supplied by OMIM, Mar 2008].
Specifications
| Phospho-MRLC2 (Ser18) | |
| Polyclonal | |
| Unconjugated | |
| MYL12B | |
| MRLC2, MYLC2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103910, 50685, 67938 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14950, Q3THE2, P18666 | |
| MYL12B | |
| A synthetic phosphopeptide derived from human Myosin regulatory light chain 2 around the phosphorylation site of Ser18 (A-T-SP-N-V) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction