missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MLK1/MLK2 (Thr312, Thr266) Polyclonal Antibody

Description
A suggested positive control for IHC is human brain tissue.
MLK is a member of the serine/threonine kinase family. This kinase has been shown to activate MAPK8/JNK and MKK4/SEK1, and this kinase itself can be phoshorylated, and thus activated by JNK kinases. This kinase functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis.
Specifications
Specifications
| Antigen | Phospho-MLK1/MLK2 (Thr312, Thr266) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MAP3K9 |
| Gene Accession No. | P80192, Q02779, Q3U1V8, Q66L42 |
| Gene Alias | BC028668; BC046514; E130314H24Rik; Map3k10; Map3k9; MEKK10; MEKK9; mitogen activated protein kinase kinase kinase 9; mitogen-activated protein kinase kinase kinase 10; Mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity); mixed lineage kinase 2; MKN28 derived nonreceptor_type; MKN28 derived nonreceptor_type serine/threonine kinase; MKN28 kinase; MLK1; MLK2; MST; PRKE1; protein kinase MST; serine/threonine kinase |
| Gene Symbols | MAP3K10, MAP3K9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?