missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MER/SKY (Tyr749, Tyr681) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15516961
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15516961 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15516961 Supplier Invitrogen™ Supplier No. PA540270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

Receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including LGALS3, TUB, TULP1 or GAS6. Regulates many physiological processes including cell survival, migration, differentiation, and phagocytosis of apoptotic cells (efferocytosis). Ligand binding at the cell surface induces autophosphorylation of MERTK on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by ligand, interacts with GRB2 or PLCG2 and induces phosphorylation of MAPK1, MAPK2, FAK/PTK2 or RAC1. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganization and engulfment. Functions in the retinal pigment epithelium (RPE) as a regulator of rod outer segments fragments phagocytosis. Plays also an important role in inhibition of Toll-like receptors (TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MER/SKY (Tyr749, Tyr681)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene MERTK
Gene Accession No. P55144, P55146, P57097, Q06418, Q12866, Q60805
Gene Alias AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; c-Eyk; c-mer; c-mer proto-oncogene tyrosine kinase; Dtk; EC 2.7.10.2; Etk-2; etk2/tyro3; Eyk; kinase Syk; KSYK; Mer; MER proto-oncogene, tyrosine kinase; MER receptor tyrosine kinase; MERTK; nmf12; Nyk; Proto-oncogene c-Mer; proto-oncogene tyrosine-protein kinase MER; rdy; Receptor tyrosine kinase MerTK; Receptor tyrosine tinase gene probably the gene for Rdy; Rek; retinal dystrophy; RP38; RSE; Sky; sMER; sMERTK; soluble MER; soluble MERTK; STK kinase; SYK; Tif; TK19-2; Tyro 12; Tyro12; Tyro3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase Mer; Tyrosine-protein kinase receptor TYRO3; tyrosine-protein kinase RSE; Tyrosine-protein kinase SKY; tyrosine-protein kinase TIF
Gene Symbols MERTK, TYRO3
Host Species Rabbit
Immunogen Synthesized peptide derived from human MerTK/Tyro3 around the phosphorylation site of Y749/681.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10461, 17289, 22174, 25232, 65037, 7301
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.