missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MEK6 (Ser207) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536669
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 37 kDa. Purity is >95% by SDS-PAGE.
MKK6 (MEK6, MAP2K6) is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis. Tissue specificity: Isoform 2 is only expressed in skeletal muscle. Isoform 1, on the other hand, is found in skeletal muscle, heart, and in lesser extent in liver or pancreas.
Specifications
| Phospho-MEK6 (Ser207) | |
| Polyclonal | |
| Unconjugated | |
| MAP2K6 | |
| Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 6; MAP2K6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 6; MKK3; MKK6; PRKMK3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK3; Stress-activated protein kinase kinase 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114495, 26399, 5608 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P52564, P70236 | |
| MAP2K6 | |
| Synthetic phosphopeptide derived from human MEK-6 around the phosphorylation site of Serine 207. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction