missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Phospho-MEK2 (Thr394) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-MEK2 (Thr394). This antibody recognizes human, mouse antigen. The Phospho-MEK2 (Thr394) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | MEK2 (Thr394) |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 750 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | FLJ26075; MAPKK2; MEK2; MKK2; PRKMK2 |
| Host Species | Rabbit |
| Immunogen | Peptide |
| Purification Method | Antigen affinity purification |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?