missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MEK1 (Thr292) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£125.00 - £280.00

Specifications

Antigen Phospho-MEK1 (Thr292)
Concentration 450 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17291416
View Documents
Proteintech
28930-1-AP-20UL
20 μL
£125.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17281416
View Documents
Proteintech
28930-1-AP-100UL
100 μL
£280.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MEK1 is also named as MAP2K1 (mitogen-activated protein kinase kinase 1), MAPKK1, PRKMK1, MKK1 and belongs to the MAP kinase kinase subfamily. It is a dual-specificity kinase that mediate ERK1 and ERK2 activation during adhesion and growth factor signaling(PMID:19219045). It also plays an essential role in extra-embryonic ectoderm during placentogenesis. MEK1 has 2 isoforms produced by alternative splicing with the molecular weight of 43 kDa and 41 kDa. MEK1 may form a heterodimer with Mek2.(PMID:21144847,19219045).

Specifications

Specifications

Phospho-MEK1 (Thr292)
Western Blot, ELISA
Unconjugated
Rabbit
5604
450 μg/mL
Polyclonal
Liquid
Human
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.