missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MEK1 (Thr292) Mouse Monoclonal Antibody, Proteintech

Product Code. p-200106564 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30238126 20 μL 20µL
30238137 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30238126 Supplier Proteintech Supplier No. 678731IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Phospho-MEK1 (Thr292) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-MEK1 (Thr292). This antibody recognizes human, mouse, rat antigen. The Phospho-MEK1 (Thr292) antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.

Specifications

Antigen MEK1 (Thr292)
Applications Western Blot, ELISA, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 2D7A8
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols MAPKK1; MEK1; MKK1; PRKMK1
Host Species Mouse
Immunogen Peptide
Purification Method Protein G purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 40-50 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.