missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MEF2A (Thr319) Polyclonal Antibody
Phospho-MEF2A (Thr319) Polyclonal Antibody
Brand: Invitrogen PA536667
This item is not returnable.
View return policy
Description
Phospho-MEF2A (Thr319) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Phospho-MEF2A (Thr319) | |
| Polyclonal | |
| Unconjugated | |
| MEF2A | |
| A430079H05Rik; ADCAD1; C5DELq14.3; DEL5q14.3; MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); MADS box transcription enhancer factor 2, polypeptide C; mef2; Mef2a; MEF2C; myocyte enhancer factor 2a; myocyte enhancer factor 2C; Myocyte-specific enhancer factor 2A; myocyte-specific enhancer factor 2C; RSRFC4; RSRFC9; Serum response factor-like protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17258, 309957, 4205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| Q02078, Q2MJT0, Q60929 | |
| MEF2A | |
| Synthetic phosphopeptide derived from human MEF-2 around the phosphorylation site of Threonine 319. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction