missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MBP (Tyr203) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA512985
This item is not returnable.
View return policy
Description
Phospho-MBP (Tyr203) Polyclonal Antibody for DB
The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called Golli-MBP) that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes.
Specifications
| Phospho-MBP (Tyr203) | |
| Polyclonal | |
| Unconjugated | |
| MBP | |
| 20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 4155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02686 | |
| MBP | |
| KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y203 of human MBP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction