missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MARK Pan (Thr215) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16672255
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16672255 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16672255 Supplier Invitrogen™ Supplier No. PA5105814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MARK1/2/3/4 only when phosphorylated at Thr215.

MARK1 (MAP/microtubule affinity-regulating kinase 1) belongs to the MARK family of serine/threonine kinases. MARK family protein kinases phosphorylate microtubule-associated proteins and may play a role in cytoskeletal stability. MARK2 refers to MAP/microtubule affinity-regulating kinase 2 isoform a [Homo sapiens]. EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1 called MARK2. MARK3 was originally identified as a marker that was induced by treatment with DNA damaging agents, and loss of MARK3 was found with carcinogenesis in the pancreas. MARK4 contains an N terminal serine/threonine kinase domain, a central ubiquitin associated domain, and a C terminal KA1 associated kinase domain. RT PCR analysis detects upregulated expression of the gene for MARK4 in nearly all clinical hepatocellular carcinoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MARK Pan (Thr215)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MARK3
Gene Accession No. O08678, O08679, P27448, Q03141, Q05512, Q7KZI7, Q8CIP4, Q8VHF0, Q8VHJ5, Q96L34, Q9P0L2
Gene Alias 1600015G02Rik; 2410090P21Rik; A430080F22Rik; AW491150; B930025N23Rik; C79806; Cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk; EMK1; EMK-1; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; Emk3; ETK1; ETK-1; KIAA1477; Kiaa1860; KP78; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity regulating kinase 2; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MAP/microtubule affinity-regulating kinase 3; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; MARK; MARK1; mark1 {ECO:0000312; MARK2; mark2 {ECO:0000312; MARK3; MARK4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 1; microtubule affinity regulating kinase 2; microtubule affinity regulating kinase 3; microtubule affinity regulating kinase 4; mKIAA1477; mPar-1c; MPK10; MPK-10; Par-1; PAR1 homolog; PAR1 homolog b; PAR1 homolog c; PAR1A; Par-1a; Par1b; Par-1b; Par1c; Par-1c; PAR-1D; protein kinase STK10; RGD:619882}; RGD:708483}; Ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase MARK1; Serine/threonine-protein kinase MARK2; serine/threonine-protein kinase p78; testicular tissue protein Li 117
Gene Symbols MARK1, MARK2, MARK3, MARK4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MARK1(Accession Q9P0L2), corresponding to amino acid residues around phosphorylated Thr215.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117016, 13728, 170577, 17169, 2011, 226778, 232944, 4139, 4140, 57787, 60328, 680407
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.