missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517495
This item is not returnable.
View return policy
Description
This antibody does not react with MARK4. It is not recommended to aliquot this antibody.
Specifications
| Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) | |
| Polyclonal | |
| Unconjugated | |
| MARK3 | |
| 1600015G02Rik; A430080F22Rik; AW491150; B930025N23Rik; cdc25C-associated protein kinase 1; CTAK1; C-TAK1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk; EMK1; EMK-1; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; Emk3; ETK1; ETK-1; Kiaa1477; KP78; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity regulating kinase 2; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MAP/microtubule affinity-regulating kinase 3; MARK; MARK1; mark1 {ECO:0000312; MARK2; mark2 {ECO:0000312; MARK3; microtubule affinity regulating kinase 1; microtubule affinity regulating kinase 2; microtubule affinity regulating kinase 3; mKIAA1477; mPar-1c; MPK10; MPK-10; Par-1; PAR1 homolog; PAR1 homolog b; PAR1 homolog c; PAR1A; Par-1a; Par1b; Par-1b; Par1c; Par-1c; Protein kinase STK10; RGD:619882}; RGD:708483}; ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase MARK1; Serine/threonine-protein kinase MARK2; serine/threonine-protein kinase p78; testicular tissue protein Li 117 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 117016, 13728, 170577, 17169, 2011, 226778, 4139, 4140, 60328 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 59 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 0.15M NaCl, 50% glycerol and no preservative; pH 7.5 | |
| O08678, O08679, P27448, Q03141, Q05512, Q7KZI7, Q8VHF0, Q8VHJ5, Q9P0L2 | |
| MARK1, MARK2, MARK3 | |
| Synthetic phosphopeptide corresponding to residues surrounding threonine 215 of human MARK1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction