missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MARCKS (Ser27) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105557
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of MARCKS only when phosphorylated at Ser27.
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
Specifications
| Phospho-MARCKS (Ser27) | |
| Polyclonal | |
| Unconjugated | |
| MARCKS | |
| 80K protein; 80K-L; 80K-L protein; FLJ14368; FLJ90045; KINC; Macs; MARCKS; MARCS; myristoylated alanine rich protein kinase C substrate; myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein; protein kinase C substrate, 80 kDa protein, light chain | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 17118, 25603, 4082 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26645, P29966, P30009 | |
| MARCKS | |
| A synthesized peptide derived from human MARCKS(Accession P29966), corresponding to amino acid residues around phosphorylated Ser27. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction