missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MARCKS (Ser167, Ser170) Recombinant Rabbit Monoclonal Antibody (MARCKSS167170-C9), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£184.00 - £464.00
Spécification
| Antigen | Phospho-MARCKS (Ser167, Ser170) |
|---|---|
| Clone | MARCKSS167170-C9 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | APC |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
17157028
|
Invitrogen™
MA537439 |
10 Tests |
£184.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17176778
![]() |
Invitrogen™
MA537191 |
100 Tests |
£464.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.Spécification
| Phospho-MARCKS (Ser167, Ser170) | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Human, Mouse, Rat | |
| P26645, P29966, P30009 | |
| 17118, 25603, 4082 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser167/170 of human phospho MARCKS. | |
| Primary | |
| 4°C | |
| MARCKS |
| MARCKSS167170-C9 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 80K protein; 80K-L; 80K-L protein; FLJ14368; FLJ90045; KINC; Macs; MARCKS; MARCS; myristoylated alanine rich protein kinase C substrate; myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein; protein kinase C substrate, 80 kDa protein, light chain | |
| MARCKS | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit