missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MARCKS (Ser163) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538139
This item is not returnable.
View return policy
Description
Myristoylated alanine-rich protein kinase C substrate (MARCKS), also designated 80K or 80K-L, has been identified as a major cellular substrate for protein kinase C. Human MARCKS is a 332 amino acid protein with a calculated molecular weight of 31.534 kDa; however, it has been shown to run at 80-87 kDa on Western blot. The plasma membrane bound protein dissociates from the membrane upon phosphorylation by various PKC isoforms. In NIH/3T3 fibroblasts, PKC alpha and PKC epsilon, but not PKC delta, are responsible for MARCKS phosphorylation. MARCKS has been found to bind calmodulin, Actin and Synapsin and is a filamentous (F) Actin crosslinking protein.
Specifications
| Phospho-MARCKS (Ser163) | |
| Polyclonal | |
| Unconjugated | |
| MARCKS | |
| 80K-L protein, MACS, MARCS, Myristoylated alanine-rich C-kinase substrate, PKCSL, PRKCSL, Protein kinase C substrate, 81 kDa protein, light chain | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17118, 25603, 4082 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29966, P26645, P30009 | |
| MARCKS | |
| A synthetic phosphopeptide derived from human MARCKS around the phosphorylation site of Ser162 (K-K-SP-F-K) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction