missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MAPKAPK2 (Thr334) Recombinant Rabbit Monoclonal Antibody (MAPKAPK2T334-H2), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537011
This item is not returnable.
View return policy
Description
MAPKAPK2 mediates p38 and ERK signaling in vivo. It is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation, and cell proliferation. MAPKAPK2 encodes a member of the Ser/Thr protein kinase family. MAPKAPK2 is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, MAPKAPK2 is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for MAPKAPK2.
Specifications
| Phospho-MAPKAPK2 (Thr334) | |
| Recombinant Monoclonal | |
| PE | |
| MAPKAPK2 | |
| AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 17164, 9261 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| MAPKAPK2T334-H2 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P49137, P49138 | |
| MAPKAPK2 | |
| A synthetic phospho-peptide corresponding to residues surrounding Thr334 of human phospho MAPKAPK-2. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction