missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MAPK9 (Thr183, Tyr185) Recombinant Rabbit Monoclonal Antibody (SAPKT183Y185-A11), FITC, Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£185.00 - £463.00

Specifications

Antigen Phospho-MAPK9 (Thr183, Tyr185)
Clone SAPKT183Y185-A11
Applications Flow Cytometry
Classification Recombinant Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17166968
View Documents
Invitrogen™
MA537380
10 Tests
£185.00
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17106728
GREENER_CHOICE
View Documents
Invitrogen™
MA537134
100 Tests
£463.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
Specifications

Specifications

Phospho-MAPK9 (Thr183, Tyr185)
Flow Cytometry
FITC
Rabbit
Human
P45984
5601
A synthetic phospho-peptide corresponding to residues surrounding Thr183/Tyr185 of human phospho SAPK/JNK.
Primary
4°C, store in dark
MAPK9
SAPKT183Y185-A11
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK/SAPK alpha; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM8; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
MAPK9
IgG κ
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.