missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MAP2K4 (Thr261) Polyclonal Antibody

Phospho-MAP2K4 (Thr261) Polyclonal Antibody
Brand: Invitrogen PA5104909
This item is not returnable.
View return policy
Description
Phospho-MAP2K4 (Thr261) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Phospho-MAP2K4 (Thr261) | |
| Polyclonal | |
| Unconjugated | |
| MAP2K4 | |
| C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 26398, 287398, 6416 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P45985, P47809 | |
| MAP2K4 | |
| A synthesized peptide derived from human MAP2K4(Accession P45985), corresponding to amino acid residues around phosphorylated Thr261. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction